Learn More
GDF-9 Antibody (53/1), DyLight 755, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP261934IR
Catalog No. NBP261934IR
Add to Cart
Description
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
GDF-9
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
GDF9
Tuberculin coupled peptide with sequence VPAKYSPLSVLTIEPDGSIAYKEYEDMIATKC that recognizes an epitope with the EPDG sequence near the C-terminal region of human GDF-9
0.1 mL
Primary
Human
Purified
ELISA, Immunohistochemistry, Western Blot
53/1
Western Blot, ELISA, Immunohistochemistry
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9
Mouse
Protein A purified
Cytokine Research
2661
Store at 4°C in the dark.
IgG1
Product Suggestions
Product Suggestions