Learn More
GDF-9 Mouse anti-Human, Alexa Fluor 594, Clone: 917319, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8266T100UG
Catalog No. FAB8266T100
Add to cart
This item is not returnable. View return policy
Description
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GDF-9
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9
Human embryonic kidney cell line derived recombinant human GDF-9 Met1-Arg454 Accession # O60383
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human GDF-9 in direct ELISAs.
Flow Cytometry
917319
Flow Cytometry 0.25-1 ug/10^6 cells
O60383
Mouse
IgG2b
RUO
Primary
2661
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)