Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDF-9 Antibody (72B), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP261933UV
Description
GDF-9 Monoclonal specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.Specifications
GDF-9 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
GDF9 | |
N-terminal of GDF-9 corresponding to sequence KKPLGPASFNLSEYFC | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Western Blot | |
72B | |
Western Blot, ELISA | |
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9 | |
Rat | |
Protein G purified | |
Cytokine Research | |
2661 | |
Store at 4°C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction