Learn More
GDF-9 Rat anti-Human, DyLight 550, Clone: 72B, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP261933R
Catalog No. NBP261933R
Add to cart
Description
Description
GDF-9 Monoclonal antibody specifically detects GDF-9 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
GDF-9
72B
Western Blot, ELISA
Purified
GDF9
N-terminal of GDF-9 corresponding to sequence KKPLGPASFNLSEYFC
Protein G purified
Cytokine Research
Primary
2661
ELISA, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
GDF9, GDF-9, growth differentiation factor 9, growth/differentiation factor 9
Rat
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
Human