Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GDF3 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547974
Description
In Western blots, less than 5% cross-reactivity with recombinant mouse (rm) GDF-5, rmGDF-6, rmGDF-7, rmGDF-8, and rmGDF-9 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.
Specifications
GDF3 | |
Polyclonal | |
Unconjugated | |
GDF3 | |
C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
14562, 9573 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q07104, Q9NR23 | |
GDF3 | |
E. coli-derived recombinant mouse GDF-3 Ala253-Gly366. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction