Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ GDF5 (Human) ELISA Kit
Click to view available options
Quantity:
1 Kit
Description
GDF5 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for the quantitative measurement of human GDF5
Specifications
Specifications
Description | growth differentiation factor 5 |
Cross Reactivity | Human |
Target Species | Human |
For Use With (Application) | Quantitative |
Gene ID (Entrez) | 8200 |
Quantity | 1 Kit |
Regulatory Status | RUO |
Storage Requirements | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction