Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDF5OS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549040

Catalog No. PA549040

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

GDF5OS
Polyclonal
PBS with 0.09% sodium azide
Q5U4N7
Protein GDF5OS, mitochondrial, Growth/differentiation factor 5 opposite strand transcript protein, GDF5OS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
554250
Western Blot
Unconjugated
GDF5OS
Liquid
GDF5OS
KLH conjugated synthetic peptide between 206-232 amino acids from the C-terminal region of human GDF5OS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit