Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner Glucose Dehydrogenase Activity Colorimetric Assay Kit: Glucose Dehydrogenase (GDH) Activity Assay Kit: Colorimetric Assay for Detecting GDH Activity in Cells, Tissues, Serum etc. Simple, Reliable & Sensitive. Detection Limit: 0.01 mU. 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K786100
Glucose 1-dehydrogenase (NADᶧ) (EC 1.1.1.118 ) is an enzyme that catalyzes the chemical reaction: D-glucose + NADᶧ↔D-glucono-1,5-lactone + NADH + H+. This enzyme belongs to the family of oxidoreductases, specifically those acting on the CH-OH group of donor with NADᶧ or NADPᶧ as acceptor. BioVision’s Glucose Dehydrogenase (GDH) Assay Kit provides a convenient tool for sensitive detection of the GDH in a variety of samples. The GDH present in sample will recognize D-glucose as a specific substrate leading to a proportional color development. The activity of GDH can be easily quantified colorimetrically (λ = 450 nm). This assay detects GDH activity as low as 0.01 mU with our unit definition. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9904