Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDNF Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0308-12F9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212335AF350

Catalog No. NP21233A30


Add to cart

Description

Description

GDNF Monoclonal antibody specifically detects GDNF in Human samples. It is validated for Western Blot.
Specifications

Specifications

GDNF
MM0308-12F9
Western Blot
Conjugated, Purified
GDNF
Human GDNF
Protein G purified
Apoptosis, Biologically Active Proteins, Cancer
Primary
2668.0
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit