Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GDNF Rabbit anti-Human, Clone: JA93-10, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP266866

Catalog No. NBP266866


Add to Cart

Description

Description

GDNF Monoclonal specifically detects GDNF in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

GDNF
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Rabbit
Protein A purified
Apoptosis, Biologically Active Proteins, Cancer
2668
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
JA93-10
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
GDNF
Synthetic peptide within human GDNF aa 51-100/211. (SwissProt: P39905 Human; SwissProt: P48540 Mouse; SwissProt: Q07731 Rat)
100 μL
Primary
Human, Mouse, Rat
Purified
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit