Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDPD1/GDE4 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13325R

Description
Hydrolyzes lysoglycerophospholipids to produce lysophosphatidic acid (LPA) and the corresponding amines. Shows a preference for 1-O-alkyl-sn-glycero-3-phosphocholine (lyso-PAF), lysophosphatidylethanolamine (lyso-PE) and lysophosphatidylcholine (lyso-PC). May be involved in bioactive N-acylethanolamine biosynthesis. Does not display glycerophosphodiester phosphodiesterase activity, since it cannot hydrolyze either glycerophosphoinositol or glycerophosphocholine.Specifications
GDPD1/GDE4 | |
Polyclonal | |
Unconjugated | |
Gdpd1 | |
2610020H15Rik; BOS_18359; Gde4; GDPD1; glycerophosphodiester phosphodiesterase 1-like; Glycerophosphodiester phosphodiesterase 4; glycerophosphodiester phosphodiesterase domain containing 1; Glycerophosphodiester phosphodiesterase domain-containing protein 1; LOC100352793; Lysophospholipase D GDPD1; RGD1311813 | |
Rabbit | |
Protein A | |
RUO | |
284161 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q8N9F7 | |
Gdpd1 | |
KLH conjugated synthetic peptide derived from human GDPD1/GDE4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction