Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDPD3 Rabbit anti-Human, DyLight 594, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297884DL594
Description
GDPD3 Polyclonal specifically detects GDPD3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
GDPD3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
EC 3.1, FLJ22603, glycerophosphodiester phosphodiesterase domain containing 3, glycerophosphodiester phosphodiesterase domain-containing protein 3, MGC4171 | |
Produced in rabbits immunized with E. coli-derived Human GDPD3 fragment. | |
0.1 mL | |
Lipid and Metabolism | |
79153 | |
Store at 4°C in the dark. | |
IgG |
Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title