Learn More
GDPGP1/C15orf58 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS13326R
Encompass_Preferred
Catalog No. 50-198-2806
Add to cart
Description
Description
Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.
Specifications
Specifications
GDPGP1/C15orf58
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GDPGP1
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
390637
GDPGP1
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Q6ZNW5
C15orf58; D330012F22Rik; GDP-D-glucose phosphorylase 1; GDP-D-glucose phosphorylase C15orf58; GDP-D-glucose phosphorylase C15orf58 homolog; GDPGP1; VTC2
KLH conjugated synthetic peptide derived from human GDPGP1/C15orf58.
Protein A
RUO
Antibody
Polyclonal
Human