Learn More
GE Healthcare 2'5' ADP Sepharose™ 4B medium
2'5' ADP is an NADP structural analog immobilized on Sepharose 4B
Manufacturer:  GE Healthcare 17070001
Catalog No. 45-000-133
Add to cart
Description
Description
  • 2'5' ADP Sepharose 4B binds and immobilizes enzymes requiring NADP cofactor
Specifications
Specifications
>0.4mg G6PDH/mL
Used for purification of Enzymes requiring NADP as a cofactor
4% agarose
90μm
5g
Approx. 2 μmol 2'5' ADP/mL drained medium
2μM 2′5′ ADP/mL Drained Medium
4% Agarose
4 to 10
2° to 8°C