Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Quantitative precipitation separates proteins from detergents, salts, lipids, phenolics and nucleic acids
- Does not result in spot gain or loss
- Processes 50 samples of up to 100μL each, but is scaleable for larger volumes or more diliute samples
- Delivers yield in about 90 minutes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Boiling Point | 1600°C |
| Color | Colorless |
| Product Type | Electrophoresis Clean-up Kit |
| Form | Liquid |
| For Use With (Application) | Improves quality of 2-D electrophoresis |
| Kit Contents | 15mL Precipitant, 17.5mL coprecipitant, 50mL wash Buffer, 250μL wash additive, quick reference card, instructions |
| Melting Temperature | 851°C |
| Molecular Weight (g/mol) | 105.98874 |
| Storage Requirements | −20°C |
| Sufficient For | 50 samples of less than 100μL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
