Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare 2-D Protein Extraction Buffer Trial Kit

Provides a convenient way to prepare high-quality protein lysates

Manufacturer:  GE Healthcare 28943522

Catalog No. 45-002-168


Add to cart

Includes:
  • 2-D Protein extraction buffer-I (Urea, NP-40)
  • 2-D Protein extraction buffer-II (Urea, CHAPS)
  • 2-D Protein extraction buffer-III (Urea, Thiourea, ASB-16, CHAPS)
  • 2-D Protein extraction buffer-IV (Urea, Thiourea, SB 3-10, CHAPS)
  • 2-D Protein extraction buffer-V (Urea, Thiourea, CHAPS)
  • 2-D Protein extraction buffer-VI (Urea, Thiourea, NDSB 201, CHAPS)
  • One 7mL DILUENT-I
  • One 14mL DILUENT-II
  • Two 12mL DILUENT-III

Description

Description

Efficient cell lysis and protein extraction are main steps for achieving high-quality results in downstream applications.

  • Buffers are designed to produce high spot resolution for 2-D gel analysis
  • Six buffers are available as dry powder, which is rehydrated with the provided DILUENT
  • Optimal buffer will depend on sample nature
  • Allows to evaluate each extraction buffer to find the most suitable buffer for sample

Order Info

Complete packsize: 50mL

Specifications

Specifications

7 mL Diluent-I, 14 mL Diluent-II, 2 x 12 mL Diluent-III
Solid and Liquid
2° to 8°C
White
  • 2-D Protein extraction buffer-I (Urea, NP-40)
  • 2-D Protein extraction buffer-II (Urea, CHAPS)
  • 2-D Protein extraction buffer-III (Urea, Thiourea, ASB-16, CHAPS)
  • 2-D Protein extraction buffer-IV (Urea, Thiourea, SB 3-10, CHAPS)
  • 2-D Protein extraction buffer-V (Urea, Thiourea, CHAPS)
  • 2-D Protein extraction buffer-VI (Urea, Thiourea, NDSB 201, CHAPS)
  • One 7mL DILUENT-I
  • One 14mL DILUENT-II
  • Two 12mL DILUENT-III
2-D Protein Extraction Buffer
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit