Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare 2-D Protein Extraction Buffers

Designed to produce high spot resolution for 2-D gel analysis

Manufacturer:  GE Healthcare 28943523

 View more versions of this product

Catalog No. 45-002-167

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Convenient way to prepare quality protein lysates
  • Ready to mix buffers simplifies protein extraction preparation of cells and tissues
  • Efficient and reproducible total protein extraction with yield
  • Can also be used prior to 1-D electrophoresis, or for rehydrating IPG strips to 2-D electrophoresis
  • Each package contains sufficient amounts of reagents to prepare 50mL buffer

Order Info

Buffer is supplied as a dry powder that is rehydrated with the provided diluent.

Specifications

Specifications

50 mL
1X
Contains <10M Urea and <10% NP-40
Buffer I with Diluent I
White
Solid
15° to 30°C
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit