GE Healthcare 8-Well Comb for HE 33 Mini Submarine Unit, 1.5 mm Available on GSA/VA Contract for Federal Government customers only.
Well forming combs for the HE 33 horizontal Mini submarine gel electrophoresis unit
Manufacturer: GE Healthcare 80605093
 View more versions of this product
Catalog No. 45-001-844
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Thickness (Metric) 0.15cm
    Product Type Backless Comb
    For Use With (Equipment) HE33 mini submarine unit
    No. of Wells 12
    Thickness (English) 0.05 in.
  • Description & Specifications
Related Products
Product Content Feedback