Learn More
GE Healthcare Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
Manufacturer:  GE Healthcare 18101044
 View more versions of this product
Catalog No. 45-500-113
  Check Availability
Add to cart
Specifications
Description
  • Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
  • Efficient and reproducible transfers improve blotting results
  • Carry out all pretransfer steps in the unit
  • Precise vacuum control for optimal transfer conditions
  • RNA transfers in less than 4 hours
Description & Specifications
Specifications
Amersham™ VacuGene™ XL vacuum blotting system