GE Healthcare Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System Available on GSA/VA Contract for Federal Government customers only.
VacuGene XL vacuum blotting system is the choice for nucleic acid blotting.
Manufacturer: GE Healthcare 18101043
 View more versions of this product
Catalog No. 45-000-419
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Amersham™ VacuGene™ XL vacuum blotting system,Amersham™ VacuGene™ XL vacuum blotting system
    Product Type Gel Supporting Screen
    • Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
    • Efficient and reproducible transfers improve blotting results
    • Carry out all pretransfer steps in the unit
    • Precise vacuum control for optimal transfer conditions
    • RNA transfers in less than 4 hours
  • Description & Specifications
Related Products
Product Content Feedback