GE Healthcare Accessories for SE 250/260 Mini-vertical Gel Units: Spacer Available on GSA/VA Contract for Federal Government customers only.

Collection of spare part accessories for SE 250/260 Gel Units

Manufacturer: GE Healthcare 80613814

 View more versions of this product

Catalog No. 45-001-684

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) SE 250, SE 260 mini format vertical gel electrophoresis unit
    Product Type Spacer
    Length (Metric) 8cm
    Thickness (English) 0.03 in.
    Thickness (Metric) 0.1cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit