Learn More
GE Healthcare Activated Thiol Sepharose™ 4B Medium
For reversible coupling of proteins to Sepharose 4B via a glutathione spacer arm
Manufacturer:  GE Healthcare 17064001
Catalog No. 45-000-122
Add to cart
Description
Description
  • Gel reacts with heavy metal ions and with alkyl and aryl halides
  • Addition reactions occur with compounds containing C=O, C=C, and N=N bonds
  • Less substituted but more porous than Thiopropyl Sepharose 6B
  • Suitable for covalent chromatography of large molecules such as enzymes and nucleic acids
  • Possible to couple proteins lacking thiol groups to thiol gels using the heterobifunctional reagent SPDP, facilitating use of covalent chromatography
Specifications
Specifications
Used for reversible immobilization of molecules containing Thiol groups under mild conditions, optimized for immobilization of large molecules
4% agarose
90μm
15g
1μM Activated Thiol/mL Medium
4% Agarose
2 to 8
2° to 8°C