GE Healthcare Agarose NA Media Available on GSA/VA Contract for Federal Government customers only.

Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. GE Healthcare™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.

Manufacturer: GE Healthcare 17055401

 View more versions of this product

Catalog No. 45-000-099

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material Agarose,Agarose
    Color White,White
    Melting Temperature 60° to 90°C,60° to 90°C
    Quantity 10g
    For Use With (Application) DNA/RNA application,DNA/RNA application
    Format Solid,Solid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit