GE Healthcare™ Agarose NA Media Available on GSA/VA Contract for Federal Government customers only.

Highly purified agarose for separation of nucleic acids to be used as substrates for enzymes. GE Healthcare™ Agarose NA Media with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.

Manufacturer: GE Healthcare™ 17055403

 View more versions of this product

Catalog No. 45-000-101

  Check Availability
Add to Cart

    Specifications

    Material Agarose,Agarose
    Color White,White
    Melting Temperature 60° to 90°C,60° to 90°C
    Quantity 1kg
    For Use With (Application) DNA/RNA application,DNA/RNA application
    Format Solid,Solid

    • Gelling temp 34 to 37°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit