Learn More
GE Healthcare Amdex Streptavidin-Alkaline Phosphatase Enzyme Conjugate
For ELISA signal amplification
Manufacturer:  GE Healthcare RPN4402
Catalog No. 45-001-176
Add to cart
Description
Description
The conjugates are based on novel chemistry which facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone.
  • 10- to 100-fold increase in signal intensity
  • Compatible with traditional ELISA systems
  • Option to reduce incubation times
  • Well controlled nonspecific binding properties
Specifications
Specifications
0.1 M NaCl, 0.05 M Tris/HCl pH7.2, 1mM MgCl2 0.1mM ZnCl2, 15 mM NaN3, 1 mg BSA/ml
For use in ELISA systems
Streptavidin Alkaline Phosphatase
500 μl
appro. 1 mg alkaline phosphatase/ml
Liquid
0.5mL
2 - 8°C