Learn More
GE Healthcare Amerlex-M Donkey Anti-Rabbit Reagent
Amerlex-M donkey anti-rabbit reagent for use in immunoassays where the primary antibody is of rabbit origin, combining the advantages of second antibody solid phase separation with liquid phase kinetics using magnetic beads
Manufacturer:  GE Healthcare RPN510
Catalog No. 45-000-941
Add to cart
Description
Description
  • Magnetizable polymer beads coated with secondary antibody enable convenient separation of free and bound fractions eliminating the need for centrifugation
  • Particularly useful when the tracer is iodine-125
  • Reduces tube handling for minimal hands-on tme
  • Sufficient for 400 tubes in a typical assay
  • Designed for use in a magnetic separation protocol, but can be adapted for conventional centrifugation (protocol supplied)
Specifications
Specifications
Immunoassay
Amerlex-M Donkey Anti-Rabbit reagent
2° to 4°C
Liquid
220mL
400 Tubes
Safety and Handling
Safety and Handling
ShelfLife : 6 Months