GE Healthcare Amersham™ AlkPhos Direct™ Labeling and Detection System with CDP-Star

Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes

Manufacturer: GE Healthcare RPN3690

 View more versions of this product

Catalog No. 45-001-775

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Storage Conditions 2° to 8°C
    Sufficient For 2500cm2 membrane
    For Use With (Application) All high-sensitivity applications using DNA, RNA, or oligonucleotide probes
    Includes Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star detection reagents
    Quantity Each

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit