GE Healthcare Amersham™ AlkPhos Direct™ Labeling Modules Available on GSA/VA Contract for Federal Government customers only.

Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes. AlkPhos Direct Labeling Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.

Manufacturer: GE Healthcare RPN3681

 View more versions of this product

Catalog No. 45-001-774

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Storage Conditions 2° to 8°C
    Sufficient For 5000cm2 membrane
    For Use With (Application) Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
    Quantity Each
    Includes Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent

    DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.

    • Hybridization stringency can be controlled by elevating temperatures up to 75°C
    • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
    • Rapid protocol saves two to three hours over conventional indirect methods

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit