Learn More
GE Healthcare Amersham™ Amplify™ Fluorographic Reagent Available on GSA/VA Contract for Federal Government customers only.
Fast and convenient fluorographic reagent for 35S-, 14C- and 3H-labeled samples in gels
Manufacturer:  GE Healthcare NAMP100
Catalog No. 45-000-686
Add to cart
Description
Description
Increases sensitivity of detection and significantly reduce exposure times required for weak beta emitters.
  • Increased detection efficiency 1000-fold for 3H and 15-fold for 35S and 14C
  • Less than two hours total preparation time
  • Can be used with acrylamide or agarose gels with no change in protocols
  • Odorless, inorganic and fully compatible with Sensitize™ Preflash unit and Hyperfilm™ MP
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Fast and convenient fluorographic reagent for 35S-, 14C- and 3H-labeled samples in Gels
3.5 to 4.5
1000mL