Learn More
GE Healthcare Amersham™ CyDye™ Antibody Labeling Kits
Convenient way to tag antibodies with CyDye fluorescent dyes
Manufacturer:  GE Healthcare PA32000
 View more versions of this product
Catalog No. 45-000-722
Add to cart
Description
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
  • Conjugations are carried out under mild conditions and are complete within 1 hour
  • Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
Specifications
Specifications
Yellow
Cy2 Reactive Dye
Cy2 Ab Labeling Kit
Each
Cy2 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water
For the conjugation of proteins with Cy2 Reactive Dye
7 to 7.3
2 - 8°C, protected from light
896.95