GE Healthcare Amersham™ CyDye™ Antibody Labeling Kits

Convenient way to tag antibodies with CyDye fluorescent dyes

Manufacturer: GE Healthcare PA32001

 View more versions of this product

Catalog No. 45-500-146

  Check Availability
Add to Cart

    Specifications

    Product Type Cy2 mAb Labeling Kit
    Storage Requirements 2° to 8°C
    Quantity Each
    For Use With (Application) For the conjugation of Proteins with Cy2 mono-functional dye
    Dye Type Cy2 mono-functional dye
    pH Range 7 to 7.3

    CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.

    • Conjugations are carried out under mild conditions and are complete within 1 hour
    • Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings

    Order Info

    Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit