GE Healthcare Amersham™ CyDye™ Antibody Labeling Kits Available on GSA/VA Contract for Federal Government customers only.

Convenient way to tag antibodies with CyDye fluorescent dyes

Manufacturer: GE Healthcare PA33001

 View more versions of this product

Catalog No. 45-000-725

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Cy3 MAb Labeling Kit
    Storage Requirements 2° to 8°C
    Quantity Each
    For Use With (Application) For the conjugation of Proteins with Cy3 mono-functional dye
    Dye Type Cy3 mono-functional dye
    Includes All reagents required for 2 x 0.1mg reactions
    pH Range 7 to 7.3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit