GE Healthcare Amersham™ CyDye™ Post-Labeling Reactive Dye Packs

Reactive CyDye fluors specifically optimized for nucleic acid microarray labeling. GE Healthcare CyDye Post-Labelling Reactive Dye Pack provides conveniently packaged, highly reactive Cy3 and Cy5 monofunctional NHS ester to label probes via the post-labeling (amino allyl) route.

Manufacturer: GE Healthcare 25801080

 View more versions of this product

Catalog No. 45-001-289

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Sufficient For One labeling reaction of up to 0.5 to 2μg of aa-cDNA or 3 to 10μg aa-RNA
    Product Type Cy5 Post-Labeling Reactive Dye Pack
    Storage Requirements -15° to -30°C
    Quantity 12 Vials
    For Use With (Application) Only reactive CyDye fluors specifically optimized for Nucleic Acid microarray labeling
    Includes 12 Vials Cy3 NHS Ester
    pH Range 8.7
    Dye Type Cy5

    • High reactivity Cy™3 and Cy™5 dyes: 40,000pmol reactive dye per vial, at >75% reactive dye content
    • Each vial is sufficient for one labeling reaction of up to 0.5-2µg of aa-cDNA or 3-10µg aa-RNA
    • Ready-to-use, individually dispensed

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit