GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs

Use for the most accurate labeling. GE Healthcare Life Sciences™ mersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies or oligonucleotides

Manufacturer: GE Healthcare PA22000

 View more versions of this product

Catalog No. 45-000-713

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) For the labeling of biological compounds with Cy2 reactive dye
    Includes Five foil packs each containing dried dye to label 1mg of Protein
    Sufficient For Labeling 1mg Protein or 30nM Amino groups
    Product Type Cy2 bis-Reactive Dye Pack
    Storage Requirements 2° to 8°C
    Quantity Each
    Dye Type Cy2 bis-reactive dye

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit