GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: GE Healthcare PA23000

Catalog No. 45-000-714

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Storage Requirements 2° to 8°C
    Sufficient For Labeling 1mg Protein or 30nM Amino groups
    Product Type Cy3 bis-Reactive Dye Pack
    For Use With (Application) For the labeling of biological compounds with Cy3 reactive dye
    Includes Five foil packs each containing dried dye to label 1mg of Protein
    Quantity Each
    Dye Type Cy2 bis-reactive dye

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit