Learn More
GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs
Use for the most accurate labeling. GE Healthcare Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.
Manufacturer:  GE Healthcare PA25000
 View more versions of this product
Catalog No. 45-000-718
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.
Description & Specifications
Specifications
For the labeling of biological compounds with Cy5 reactive dye
Five foil packs each containing dried dye to label 1mg of Protein
Cy5 bis-Reactive Dye Pack
Each
Cy5 reactive dye
Labeling 1mg Protein or 30nM Amino groups
2° to 8°C