Learn More
GE Healthcare Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester) Available on GSA/VA Contract for Federal Government customers only.
Suitable for general labeling of amine residues
Manufacturer:  GE Healthcare PA15500
 View more versions of this product
Catalog No. 45-000-707
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Suitable for general labeling of Amine residues
Cy5.5 bis-NHS Ester
5mg
Foil packs, each containing 5, 10 or 50mg dried dye
2° to 8°C