GE Healthcare Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester) Available on GSA/VA Contract for Federal Government customers only.

Suitable for general labeling of amine residues

Manufacturer: GE Healthcare PA12000

 View more versions of this product

Catalog No. 45-000-693

  Check Availability
Add to Cart

    Specifications

    Product Type Cy2 bis-NHS Ester
    Storage Requirements 2° to 8°C
    Quantity 5mg
    For Use With (Application) Suitable for general labeling of Amine residues
    Includes Foil packs, each containing 5, 10 or 50mg dried dye

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit