Learn More
GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive Hydrazide)
For the labeling of free carbonyl groups of glycoproteins and carbohydrates
Manufacturer:  GE Healthcare PA13120
 View more versions of this product
Catalog No. 45-001-191
Add to cart
Description
Description
Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoproteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.
  • Range of bulk packs available for greatest economy
  • Example protocols included with each dye pack
Specifications
Specifications
Red
Cy3
3 to 10
2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw ycles.
765.95
For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications
Foil packs, each containing 1, 5 or 25mg dried dye
Cy3 Hydrazide
5mg
Soluble in cold water and hot water