Learn More
GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)
For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides
Manufacturer:  GE Healthcare PA15631
 View more versions of this product
Catalog No. 45-001-250
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
For labeling via Thiol groups, use in steady state Fluorescence, FP and FRET applications
Foil packs, each containing 1, 5 or 25mg dried dye
Cy5.5 Maleimide
1mg
Cy5.5
3 to 10
2° to 8°C