Learn More
GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide) Available on GSA/VA Contract for Federal Government customers only.
For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides
Manufacturer:  GE Healthcare PA15636
 View more versions of this product
Catalog No. 45-001-251
Add to cart
Description
Description
The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack
Antibody labeling
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Blue (dark)
Cy5.5
3 to 10
2 - 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
1128.42
For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications
Foil packs, each containing 1, 5 or 25mg dried dye
Cy5.5 Maleimide
25 mg
Soluble in cold water and hot water