GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

Manufacturer: GE Healthcare PA13136

 View more versions of this product

Catalog No. 45-500-141

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Cy3 Maleimide
    Storage Requirements 2° to 8°C
    Quantity 25mg
    Dye Type Cy3
    Includes Foil packs, each containing 1, 5 or 25mg dried dye
    pH Range 3 to 10
    For Use With (Application) For labeling via Thiol groups, use in steady state Fluorescence, FP and FRET applications

    The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

    • Suitable for the labeling of antibodies
    • Example protocols included with each dye pack

    Applications:

    Antibody labeling

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit