Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) Available on GSA/VA Contract for Federal Government customers only.

Use for highly accurate labeling of amine groups. GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester) contain derivitized CyDyes that carry only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies, peptides, or oligonucleotides.

Manufacturer:  GE Healthcare PA15104

 View more versions of this product

Catalog No. 45-000-706


  Check Availability
Add to cart

Specifications

Description

  • Contains derivitized CyDyes that carry only one reactive group on each dye molecule
  • pH range: 3 to 10
  • DMSO tolerant
  • Photostable
  • Example protocols included with each dye pack

Description & Specifications

Specifications

For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye
Cy5 NHS Ester
10mg
Cy5
3 to 10
2° to 8°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit