GE Healthcare Amersham™ CyDye™ Value Packs (mono-Reactive NHS Ester)

Targeted and specific labeling of amine residues on antibodies, peptides, or oligonucleotides

Manufacturer: GE Healthcare PA13605

Catalog No. 45-500-142

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Cy3.5 NHS Ester
    Storage Requirements 2° to 8°C
    Quantity 5mg
    For Use With (Application) For labeling via Amine groups, CyDyes are suitable for use in applications such as Fluorescence resonance energy transfer (FRET), time resolved-Fluorescence resonance energy transfer (TR-FRET) and Fluorescence polarization (FP)
    Dye Type Cy3.5
    Includes Foil packs, each containing 1, 5, 10, 25 or 50mg dried dye
    pH Range 3 to 10

    • Contains derivitized CyDyes that carry only one reactive group on each dye molecule
    • pH range: 3 to 10
    • DMSO tolerant
    • Photostable
    • Example protocols included with each dye pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit