GE Healthcare Amersham™ ECF™ Western Blotting Reagent Pack Available on GSA/VA Contract for Federal Government customers only.
Alkaline phosphatase conjugated secondary antibodies for detection of chemifluorescent proteins immobilized on membranes
Manufacturer: GE Healthcare RPN5783
 View more versions of this product
Catalog No. 45-000-945
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    For Use With (Application) Quantification of western blotting
    Includes Anti-Rabbit Alkaline Phosphatase conjugate, membrane-blocking agent and ECF™ Substrate
    Quantity Each,Each
    Kit Contents Anti-Rabbit Alkaline Phosphatase conjugate, membrane-blocking agent and ECF™ Substrate
  • Description & Specifications
Related Products
Product Content Feedback