GE Healthcare Amersham™ ECL™ Precast Gels

Use precast gels to save time and obtain high quality results. Amersham™ ECL™ Precast Gels are convenient to handle and perfect for downstream detection steps.

Manufacturer: GE Healthcare 28989805

 View more versions of this product

Catalog No. 45-002-462

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Shelf Life 12 months
    Format 10-well,10-well, precast Gel
    Boiling Point 219° to 220°C
    Height (Metric) 7.5cm
    Buffer Tris HCl
    Melting Temperature 168.5°C
    Molecular Weight 121.13
    Storage Requirements 4° to 8°C
    Thickness (Metric) 0.14cm
    Width (English) Gel 3.15 in.
    Width (Metric) Gel 8cm,8cm
    Formulation Acrylamide/bis-acrylamide, %C = 2.6% (w/w), %T = Total concentration (w/v) of Acrylamide + bis-acrylamide 12%
    Color Clear,Clear
    Volume Per Well 35μL
    For Use With (Application) Polyacrylamide Gel electrophoresis
    Height (English) 2.95 in.
    Concentration 12%
    Thickness (English) 0.05 in.,0.05 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit