GE Healthcare Amersham™ ECL™ Precast Gels

Manufacturer: GE Healthcare 28990156

Catalog No. 45-002-467

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Thickness (English) 0.05 in.,0.05 in.
    Thickness (Metric) 0.14cm
    Concentration 12%
    Width (English) Gel 3.15 in.
    Width (Metric) Gel 8cm,8cm
    Format 15-well, precast Gel,15-well, precast Gel
    Height (English) 2.95 in.
    Height (Metric) 7.5cm
    Buffer Tris HCl
    Color Clear,Clear
    For Use With (Application) Polyacrylamide Gel electrophoresis
    Formulation Acrylamide/bis-acrylamide, %C = 2.6% (w/w), %T = Total concentration (w/v) of Acrylamide + bis-acrylamide 12%
    Shelf Life 12 Months
    Storage Requirements 4° to 8°C
    Volume Per Well 20μL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit