Learn More
GE Healthcare Amersham™ ECL™ Precast Gels
Use precast gels to save time and obtain high quality results. Amersham™ ECL™ Precast Gels are convenient to handle and perfect for downstream detection steps.
Manufacturer:  GE Healthcare 28989806
 View more versions of this product
Catalog No. 45-002-463
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Homogeneous and gradient gels for horizontal protein electrophoresis.
  • Amersham™ ECL™ Gels have a shelf life of up to 12 months with no compromise in performance over time
  • Gels are available in 15 and 10 well formats as either homogenous (10 and 12%) or gradient (4-12%, 8-16% and 4-20%)
  • Compatible with standard Laemmli buffers
  • Works well with standard protein detection protocols
  • Amersham ECL Gel is cast without SDS, allowing the user to define the separation conditions by the composition of the running buffer and sample loading buffer
Description & Specifications
Specifications
219° to 220°C
Clear
Polyacrylamide Gel electrophoresis
Acrylamide/bis-acrylamide, %C = 2.6% (w/w), %T = Total concentration (w/v) of Acrylamide + bis-acrylamide 4 to 12%
7.5cm
121.13
0.05 in.
35μL
8cm
Tris HCl
4 to 12%
10-well
2.95 in.
168.5°C
4° to 8°C
0.14cm
3.15 in.
Safety and Handling
ShelfLife : 12 months