GE Healthcare Amersham™ ECL™ Precast Gels Available on GSA/VA Contract for Federal Government customers only.

Use precast gels to save time and obtain high quality results. Amersham™ ECL™ Precast Gels are convenient to handle and perfect for downstream detection steps.

Manufacturer: GE Healthcare 28989804

 View more versions of this product

Catalog No. 45-002-461

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Shelf Life 12 months
    Format 10-well
    Boiling Point 219° to 220°C
    Height (English) 2.95 in.
    Height (Metric) 7.5cm
    Molecular Weight 121.13
    Color Clear
    Storage Requirements 4° to 8°C
    Thickness (Metric) 0.14cm
    Volume Per Well 35μL
    Concentration 10%
    Width (English) Gel 3.15 in.
    Width (Metric) Gel 8cm
    For Use With (Application) Polyacrylamide Gel electrophoresis
    Formulation Acrylamide/bis-acrylamide, %C = 2.6% (w/w), %T = Total concentration (w/v) of Acrylamide + bis-acrylamide 10%
    Buffer Tris HCl
    Melting Temperature 168.5°C
    Thickness (English) 0.05 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit