GE Healthcare Amersham™ Hybond™ -LFP Membranes

Hydrophobic PVDF membrane with 0.2µm pore size for fluorescent Western blotting applications

Manufacturer: GE Healthcare RPN2020LFP

 View all options for this product

Catalog No. 45-001-313


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Length (Metric) 20cm
    Width (Metric) 20cm
    For Use With (Application) Protein blotting
    Material PVDF
    Binding Capacity >400μg/cm2
    Format Sheet
    Pore Size 0.2μm
    Quantity 10 Sheets

    Fluorescent detection in Western blotting offers long-lasting signal and accurate quantitation of band intensities, resulting in a broader dynamic range than film.

    • Much lower background fluorescence than other PVDF membranes, resulting in higher sensitivity
    • Compatible with a variety of blocking agents, transfer buffers and total protein blot stains
    • Suitable for protein transfer
    • Typical protein binding capacity is >400µg/cm2
    • 8 x 7.5cm membrane matches Amersham™ECL™ precast gels and can be used with the Amersham ECL Gel horizontal electrophoresis system

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit